siRNA Details
| Gene Symbol: Nr1h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163504.1 | NP_001156976.1 | 3 | 434 | ||
| NM_001163700.1 | NP_001157172.1 | 2 | 230 | |||
| NM_009108.2 | NP_033134.2 | 2 | 230 | |||
| GenBank mRNA | AK002513.1 | - | 2 | 217 | ||
| AK050566.1 | - | 2 | 211 | |||
| AK132255.1 | - | 2 | 222 | |||
| BC015261.1 | - | 2 | 201 | 4241705 | ||
| U09416.1 | - | 2 | 199 | |||
| U09417.1 | - | 3 | 398 | |||
| U09418.1 | - | 3 | 395 | |||

Nr1h4
Genomic Map