siRNA Details
| Gene Symbol: Ryk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042607.1 | NP_001036072.1 | 6 | 826 | ||
| NM_001284258.1 | NP_001271187.1 | 5 | 704 | |||
| NM_013649.3 | NP_038677.3 | 6 | 826 | |||
| GenBank mRNA | AK144355.1 | - | 5 | 644 | ||
| BC006963.1 | - | 2 | 125 | |||
| BC032275.1 | - | 6 | 560 | 5370100 | ||
| L02210.1 | - | 6 | 801 | |||
| L21707.1 | - | 6 | 836 | |||
| L38394.1 | - | 6 | 749 | |||
| M98547.1 | - | 6 | 790 | |||

Ryk
Genomic Map