siRNA Details
| Gene Symbol: S100a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011310.2 | NP_035440.1 | 3 | 243 | ||
| GenBank mRNA | AF004941.1 | - | 3 | 243 | ||
| AF087470.1 | - | 3 | 218 | |||
| AK133717.1 | - | 3 | 245 | |||
| AK161105.1 | - | 1 | 1990 | |||
| BC099893.1 | - | 3 | 240 | 40043723 | ||
| BC100690.1 | - | 3 | 240 | 40043727 | ||
| BC100696.1 | - | 3 | 240 | 40043722 | ||
| BC100697.1 | - | 3 | 240 | 40043719 | ||

S100a3
Genomic Map