siRNA Details
| Gene Symbol: Saa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009117.3 | NP_033143.1 | 4 | 480 | ||
| GenBank mRNA | AK136475.1 | - | 4 | 486 | ||
| BC052495.1 | - | 1 | 2322 | |||
| BC087933.1 | - | 4 | 480 | 30310171 | ||
| Gene Symbol: Saa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011314.2 | NP_035444.1 | 4 | 480 | ||
| GenBank mRNA | BC034858.1 | - | 1 | 2054 | ||
| L22190.1 | - | 3 | 478 | |||
| M11130.1 | - | 4 | 480 | |||
| U60437.1 | - | 3 | 447 | |||
| U60438.1 | - | 3 | 447 | |||

Saa1
Genomic Map