siRNA Details
| Gene Symbol: Apcs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011318.2 | NP_035448.2 | 2 | 566 | ||
| GenBank mRNA | AK144248.1 | - | 2 | 453 | ||
| AK165699.1 | - | 2 | 453 | |||
| BC061125.1 | - | 2 | 463 | 30308397 | ||
| M23552.1 | - | 2 | 542 | |||
| X14079.1 | - | 2 | 542 | |||
| Y00426.1 | - | 2 | 555 | |||

Apcs
Genomic Map