siRNA Details
| Gene Symbol: Sar1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009120.2 | NP_033146.1 | 4 | 320 | ||
| GenBank mRNA | AK157843.1 | - | 4 | 332 | ||
| AK159239.1 | - | 4 | 331 | |||
| AK168175.1 | - | 4 | 332 | |||
| BC005549.1 | - | 4 | 321 | 3585667 | ||
| L20294.1 | - | 4 | 295 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK129305.1 | - | 3518 | Chr. - - [-] on Build | ||

Sar1a
Genomic Map