siRNA Details
| Gene Symbol: Sars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204979.1 | NP_001191908.1 | 9 | 1256 | ||
| NM_011319.3 | NP_035449.1 | 9 | 1256 | |||
| GenBank mRNA | AK028358.1 | - | 9 | 1188 | ||
| AK075827.1 | - | 9 | 1187 | |||
| AK077309.1 | - | 9 | 1137 | |||
| AK082793.1 | - | 9 | 1167 | |||
| AK152150.1 | - | 9 | 1211 | |||
| AK153158.1 | - | 9 | 1206 | |||
| BC008612.1 | - | 9 | 1145 | 3257101 | ||
| BC031932.1 | - | 9 | 1146 | |||

Sars
Genomic Map