siRNA Details
| Gene Symbol: Atxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199304.1 | NP_001186233.1 | 6 | 1005 | ||
| NM_001199305.1 | NP_001186234.1 | 7 | 1130 | |||
| NM_009124.6 | NP_033150.2 | 7 | 1130 | |||
| GenBank mRNA | AK048268.1 | - | 7 | 1117 | ||
| AK169931.1 | - | 7 | 1117 | |||
| BC058178.1 | - | 6 | 971 | 5326559 | ||

Atxn1
Genomic Map