siRNA Details
| Gene Symbol: Scn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018733.2 | NP_061203.2 | 11 | 1651 | ||
| GenBank mRNA | AK134744.1 | - | 11 | 1653 | ||
| BC023034.1 | - | 3 | 454 | |||
| BC034643.1 | - | 3 | 454 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scn1a| Gene Symbol: Scn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018733.2 | NP_061203.2 | 11 | 1651 | ||
| GenBank mRNA | AK134744.1 | - | 11 | 1653 | ||
| BC023034.1 | - | 3 | 454 | |||
| BC034643.1 | - | 3 | 454 | |||