siRNA Details
| Gene Symbol: Scn5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253860.1 | NP_001240789.1 | 6 | 896 | ||
| NM_021544.4 | NP_067519.2 | 7 | 1064 | |||
| GenBank mRNA | AJ271477.1 | - | 6 | 892 | ||
| AK147254.1 | - | 7 | 1064 | |||
| AK147517.1 | - | 7 | 1064 | |||

Scn5a
Genomic Map