siRNA Details
| Gene Symbol: CXCR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168298.1 | NP_001161770.1 | 4 | 772 | ||
| NM_001557.3 | NP_001548.1 | 3 | 980 | |||
| XM_005246530.3 | XP_005246587.1 | 4 | 752 | |||
| XM_017003990.1 | XP_016859479.1 | 4 | 732 | |||
| XM_017003991.1 | XP_016859480.1 | 2 | 643 | |||
| XM_017003992.1 | XP_016859481.1 | 2 | 913 | |||
| GenBank mRNA | AK290906.1 | - | 3 | 878 | ||
| AK312664.1 | - | 3 | 871 | |||
| BC037961.1 | - | 3 | 934 | 5752441 | ||
| L19593.1 | - | 3 | 970 | |||
| M73969.1 | - | 1 | 587 | |||
| M94582.1 | - | 2 | 614 | |||

CXCR2
Genomic Map