siRNA Details
| Gene Symbol: Scnn1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001272023.1 | NP_001258952.1 | 2 | 198 | ||
| NM_011325.2 | NP_035455.1 | 2 | 195 | |||
| GenBank mRNA | AF112186.1 | - | 2 | 122 | ||
| AK028909.1 | - | 2 | 177 | |||
| AK036819.1 | - | 2 | 196 | |||
| AK087055.1 | - | 2 | 157 | |||
| AK164709.1 | - | 2 | 158 | |||
| BC131969.1 | - | 2 | 111 | 40130246 | ||
| BC132397.1 | - | 2 | 111 | 40130674 | ||

Scnn1b
Genomic Map