siRNA Details
| Gene Symbol: Scrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009136.3 | NP_033162.1 | 2 | 316 | ||
| GenBank mRNA | AJ223206.1 | - | 2 | 316 | ||
| AK046213.1 | - | 2 | 185 | |||
| BC058532.1 | - | 204 | ||||
| CT010374.1 | - | 1 | 100 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scrg1| Gene Symbol: Scrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009136.3 | NP_033162.1 | 2 | 316 | ||
| GenBank mRNA | AJ223206.1 | - | 2 | 316 | ||
| AK046213.1 | - | 2 | 185 | |||
| BC058532.1 | - | 204 | ||||
| CT010374.1 | - | 1 | 100 | |||