siRNA Details
| Gene Symbol: Ccl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011329.3 | NP_035459.1 | 1, 2 | 150 | ||
| GenBank mRNA | AF065924.1 | - | 1, 2 | 129 | ||
| AF065925.1 | - | 1, 2 | 129 | |||
| AF065926.1 | - | 1, 2 | 129 | |||
| AF065927.1 | - | 1, 2 | 129 | |||
| AF065928.1 | - | 1, 2 | 129 | |||
| AK019040.1 | - | 1, 2 | 150 | |||
| BC120806.1 | - | 1, 2 | 141 | 40129729 | ||
| M17957.1 | - | 1, 2 | 153 | |||
| M23501.1 | - | 1, 2 | 150 | |||

Ccl1
Genomic Map