siRNA Details
| Gene Symbol: Ccl25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009138.3 | NP_033164.1 | 3 | 288 | ||
| GenBank mRNA | AJ249480.1 | - | 3 | 261 | ||
| AK131875.1 | - | 3 | 289 | |||
| AK132700.1 | - | 3 | 313 | |||
| AK134138.1 | - | 3 | 273 | |||
| BC117033.1 | - | 2 | 221 | 40126352 | ||
| BC117037.1 | - | 2 | 221 | 40126356 | ||
| DQ158256.1 | - | 3 | 274 | |||
| U86357.1 | - | 2 | 170 | |||

Ccl25
Genomic Map