siRNA Details
| Gene Symbol: Cxcl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011339.2 | NP_035469.1 | 4 | 330 | ||
| GenBank mRNA | AF082859.1 | - | 4 | 293 | ||
| AK142955.1 | - | 4 | 332 | |||
| BC061138.1 | - | 4 | 323 | 30298837 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cxcl15| Gene Symbol: Cxcl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011339.2 | NP_035469.1 | 4 | 330 | ||
| GenBank mRNA | AF082859.1 | - | 4 | 293 | ||
| AK142955.1 | - | 4 | 332 | |||
| BC061138.1 | - | 4 | 323 | 30298837 | ||