siRNA Details
| Gene Symbol: Sdf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009143.3 | NP_033169.3 | 3 | 666 | ||
| GenBank mRNA | AK002496.1 | - | 3 | 422 | ||
| BC058798.1 | - | 3 | 666 | 6485217 | ||
| BC062881.1 | - | 3 | 412 | 5693309 | ||
| BC087871.1 | - | 3 | 311 | 30279332 | ||
| D50646.1 | - | 3 | 420 | |||

Sdf2
Genomic Map