siRNA Details
| Gene Symbol: Sdf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK157860.1 | - | 6 | 918 | ||
| AK167113.1 | - | 6 | 967 | |||
| AK178167.1 | - | 2 | 177 | |||
| AK207454.1 | - | 2 | 198 | |||
| AK214240.1 | - | 3 | 372 | |||
| BC005715.1 | - | 7 | 1187 | |||
| BC068152.1 | - | 6 | 1293 | 30534982 | ||
| D50461.1 | - | 6 | 1296 | |||
| U45977.1 | - | 6 | 1201 | |||
| U45978.1 | - | 6 | 1069 | |||

Sdf4
Genomic Map