siRNA Details
| Gene Symbol: Frrs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113478.1 | NP_001106950.1 | 10 | 1219 | ||
| NM_009146.3 | NP_033172.2 | 10 | 1181 | |||
| GenBank mRNA | AK144933.1 | - | 10 | 1173 | ||
| AK162693.1 | - | 10 | 1174 | |||
| AK167289.1 | - | 10 | 1209 | |||
| BC027770.1 | - | 9 | 1179 | 5344922 | ||
| D50464.1 | - | 10 | 1169 | |||

Frrs1
Genomic Map