siRNA Details
| Gene Symbol: Sec22b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011342.4 | NP_035472.1 | 3 | 376 | ||
| GenBank mRNA | AK010363.1 | - | 3 | 385 | ||
| AK017237.1 | - | 3 | 382 | |||
| AK034973.1 | - | 3 | 388 | |||
| AK037918.1 | - | 3 | 377 | |||
| AK088514.1 | - | 3 | 376 | |||
| AK089928.1 | - | 3 | 369 | |||
| AK134215.1 | - | 3 | 377 | |||
| AK178690.1 | - | 2 | 176 | |||
| BC009024.1 | - | 3 | 338 | 3481731 | ||
| U91538.1 | - | 3 | 333 | |||

Sec22b
Genomic Map