siRNA Details
| Gene Symbol: Exoc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009148.3 | NP_033174.2 | 16 | 2517 | ||
| GenBank mRNA | AF022962.1 | - | 16 | 2543 | ||
| AK028250.1 | - | 14 | 2249 | |||
| AK173235.1 | - | 16 | 2520 | |||
| AK197686.1 | - | 2 | 195 | |||
| AK206003.1 | - | 2 | 281 | |||
| BC034644.1 | - | 16 | 2517 | 5355276 | ||

Exoc4
Genomic Map