siRNA Details
| Gene Symbol: Sel1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039089.1 | NP_001034178.1 | 3 | 397 | ||
| NM_011344.2 | NP_035474.1 | 3 | 397 | |||
| GenBank mRNA | AF063095.1 | - | 3 | 346 | ||
| AK005023.2 | - | 3 | 370 | |||
| AK005544.1 | - | 3 | 380 | |||
| AK146022.1 | - | 3 | 371 | |||
| AK160420.1 | - | 3 | 379 | |||
| AK160936.1 | - | 3 | 355 | |||
| AK161546.1 | - | 3 | 371 | |||
| AK220502.1 | - | 3 | 397 | |||
| BC026816.1 | - | 3 | 369 | 4457637 | ||

Sel1l
Genomic Map