siRNA Details
| Gene Symbol: Sel1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039089.1 | NP_001034178.1 | 18 | 1927 | ||
| NM_011344.2 | NP_035474.1 | 18 | 1777 | |||
| GenBank mRNA | AF063095.1 | - | 18 | 1726 | ||
| AK005023.2 | - | 18 | 1900 | |||
| AK146022.1 | - | 18 | 1901 | |||
| AK160936.1 | - | 18 | 1735 | |||
| AK220502.1 | - | 18 | 1927 | |||
| BC026816.1 | - | 18 | 1749 | 4457637 | ||
| BC049959.1 | - | 13 | 1139 | |||
| BC057452.1 | - | 4 | 362 | |||

Sel1l
Genomic Map