siRNA Details
| Gene Symbol: Selenbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009150.3 | NP_033176.2 | 12 | 1346 | ||
| GenBank mRNA | AK002447.1 | - | 12 | 1362 | ||
| BC011202.1 | - | 12 | 1346 | 4209393 | ||
| M32032.1 | - | 1329 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Selenbp1| Gene Symbol: Selenbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009150.3 | NP_033176.2 | 12 | 1346 | ||
| GenBank mRNA | AK002447.1 | - | 12 | 1362 | ||
| BC011202.1 | - | 12 | 1346 | 4209393 | ||
| M32032.1 | - | 1329 | ||||