siRNA Details
Gene Symbol: Sema3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243072.1 | NP_001230001.1 | 11 | 1438 | ||
NM_001243073.1 | NP_001230002.1 | 11 | 1278 | |||
NM_009152.4 | NP_033178.2 | 10 | 1706 | |||
GenBank mRNA | AK140817.1 | - | 11 | 1279 | ||
BC057588.1 | - | 10 | 1353 | 6416990 | ||
BC090844.1 | - | 11 | 1438 | 30532393 | ||
D85028.2 | - | 10 | 1708 | |||
L41541.1 | - | 10 | 1057 | |||
X85993.1 | - | 10 | 1167 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_017310, XM_006235989, XM_006235990, XM_008762702, X95286.
Panther Classification:
Gene Ontology Categories:
