siRNA Details
| Gene Symbol: Sema3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042779.2 | NP_001036244.1 | 10 | 1118 | ||
| NM_001291537.1 | NP_001278466.1 | 10 | 1382 | |||
| NM_001291538.1 | NP_001278467.1 | 10 | 1376 | |||
| NM_001291539.1 | NP_001278468.1 | 9 | 1059 | |||
| NM_009153.3 | NP_033179.2 | 8 | 928 | |||
| GenBank mRNA | AK014333.1 | - | 8 | 874 | ||
| AK033597.1 | - | 9 | 1069 | |||
| AK037034.1 | - | 10 | 1107 | |||
| AK043634.1 | - | 9 | 1012 | |||
| AK048364.1 | - | 9 | 1340 | |||
| AK148232.1 | - | 10 | 1079 | |||
| BC090669.1 | - | 9 | 1039 | 30696359 | ||
| BC150758.1 | - | 7 | 774 | 9087669 | ||
| CN539668.1 | - | 5 | 461 | |||
| X85990.1 | - | 8 | 925 | |||

Sema3b
Genomic Map