siRNA Details
| Gene Symbol: Sema5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009154.2 | NP_033180.2 | 11 | 1875 | ||
| GenBank mRNA | AK031231.1 | - | 11 | 1868 | ||
| AK043386.1 | - | 11 | 1587 | |||
| AK046502.1 | - | 11 | 1651 | |||
| AK141612.1 | - | 11 | 1819 | |||
| AK143026.1 | - | 11 | 1886 | |||
| AK157706.1 | - | 11 | 1858 | |||
| BC065137.1 | - | 10 | 1344 | 30548165 | ||
| X97817.1 | - | 9 | 1244 | |||

Sema5a
Genomic Map