siRNA Details
| Gene Symbol: Sema6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018744.2 | NP_061214.2 | 19 | 3412 | ||
| GenBank mRNA | AF288666.1 | - | 19 | 2665 | ||
| BC059238.1 | - | 19 | 3374 | 6417475 | ||
| BC062979.1 | - | 18 | 2825 | 6841689 | ||

Sema6a
Genomic Map