siRNA Details
| Gene Symbol: Sept8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252332.1 | NP_001239261.1 | 3 | 482 | ||
| NM_001252333.1 | NP_001239262.1 | 3 | 482 | |||
| NM_033144.2 | NP_149156.1 | 3 | 482 | |||
| GenBank mRNA | AB093215.1 | - | 3 | 332 | ||
| AF179996.1 | - | 2 | 178 | |||
| AK144178.1 | - | 3 | 324 | |||
| BC037707.1 | - | 3 | 270 | |||
| BC049819.1 | - | 3 | 297 | |||
| BC059248.1 | - | 3 | 418 | 6400429 | ||

Sept8
Genomic Map