siRNA Details
| Gene Symbol: Serf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011353.2 | NP_035483.1 | 2 | 263 | ||
| GenBank mRNA | AF073517.1 | - | 2 | 234 | ||
| AK005773.1 | - | 2 | 178 | |||
| AK012379.1 | - | 2 | 198 | |||
| BC028510.1 | - | 2 | 216 | 1510984 | ||
| BY102586.1 | - | 2 | 272 | |||

Serf1
Genomic Map