siRNA Details
| Gene Symbol: Serf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011353.2 | NP_035483.1 | 3 | 333 | ||
| GenBank mRNA | AF073517.1 | - | 3 | 304 | ||
| AK005773.1 | - | 3 | 248 | |||
| BC028510.1 | - | 3 | 286 | 1510984 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serf1| Gene Symbol: Serf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011353.2 | NP_035483.1 | 3 | 333 | ||
| GenBank mRNA | AF073517.1 | - | 3 | 304 | ||
| AK005773.1 | - | 3 | 248 | |||
| BC028510.1 | - | 3 | 286 | 1510984 | ||