siRNA Details
| Gene Symbol: Sfrp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016687.3 | NP_057896.1 | 1, 2 | 711 | ||
| GenBank mRNA | AF117709.1 | - | 1, 2 | 726 | ||
| AK009981.1 | - | 1, 2 | 709 | |||
| AK080766.1 | - | 1, 2 | 712 | |||
| BC034853.1 | - | 1, 2 | 670 | 1364770 | ||
More Information
Other Species Targeted
Set Membership: |
|

Sfrp4
Genomic Map