siRNA Details
| Gene Symbol: Sftpc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011359.2 | NP_035489.2 | 2, 3 | 237 | ||
| GenBank mRNA | BC061137.1 | - | 2, 3 | 232 | 30300758 | |
| BY773034.1 | - | 2, 3 | 238 | |||
| CJ093163.1 | - | 2, 3 | 233 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sftpc| Gene Symbol: Sftpc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011359.2 | NP_035489.2 | 2, 3 | 237 | ||
| GenBank mRNA | BC061137.1 | - | 2, 3 | 232 | 30300758 | |
| BY773034.1 | - | 2, 3 | 238 | |||
| CJ093163.1 | - | 2, 3 | 233 | |||