siRNA Details
| Gene Symbol: Sgca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009161.4 | NP_033187.1 | 4 | 428 | ||
| GenBank mRNA | AB024920.1 | - | 4 | 428 | ||
| AF019564.1 | - | 3 | 175 | |||
| AK009015.1 | - | 3 | 191 | |||
| AK075915.1 | - | 3 | 200 | |||
| BC060287.1 | - | 3 | 187 | |||

Sgca
Genomic Map