siRNA Details
| Gene Symbol: IL12RB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258214.1 | NP_001245143.1 | 8 | 1627 | ||
| NM_001258215.1 | NP_001245144.1 | 7 | 1539 | |||
| NM_001258216.1 | NP_001245145.1 | 7 | 1539 | |||
| NM_001319233.1 | NP_001306162.1 | 8 | 1623 | |||
| NM_001559.2 | NP_001550.1 | 7 | 1539 | |||
| NR_047583.1 | 7 | 1539 | ||||
| NR_047584.1 | 7 | 1539 | ||||
| XM_005270825.2 | XP_005270882.1 | 8 | 1640 | |||
| XM_005270827.2 | XP_005270884.1 | 8 | 1636 | |||
| XM_005270828.3 | XP_005270885.1 | 7 | 1548 | |||
| XM_006710617.2 | XP_006710680.1 | 8 | 1640 | |||
| XM_011541383.2 | XP_011539685.1 | 8 | 1092 | |||
| XM_011541384.2 | XP_011539686.1 | 8 | 1640 | |||
| XM_017001202.1 | XP_016856691.1 | 8 | 1640 | |||
| XM_017001203.1 | XP_016856692.1 | 7 | 1552 | |||
| XM_017001204.1 | XP_016856693.1 | 8 | 1640 | |||
| GenBank mRNA | AK294494.1 | - | 8 | 1118 | ||
| AK316525.1 | - | 8 | 1098 | |||
| BC104772.1 | - | 6 | 949 | 8143775 | ||
| BC104774.1 | - | 6 | 949 | 8143777 | ||
| BC143248.1 | - | 6 | 949 | |||
| BC143249.1 | - | 6 | 949 | 9051751 | ||
| BC143250.1 | - | 6 | 949 | |||
| U64198.1 | - | 7 | 1539 | |||

IL12RB2
Genomic Map