siRNA Details
| Gene Symbol: Zfp106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011743.2 | NP_035873.2 | 19 | 5507 | ||
| GenBank mRNA | AF060244.1 | - | 7 | 685 | ||
| AF060245.1 | - | 6 | 530 | |||
| AK029313.1 | - | 6 | 635 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp106| Gene Symbol: Zfp106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011743.2 | NP_035873.2 | 19 | 5507 | ||
| GenBank mRNA | AF060244.1 | - | 7 | 685 | ||
| AF060245.1 | - | 6 | 530 | |||
| AK029313.1 | - | 6 | 635 | |||