siRNA Details
| Gene Symbol: Itsn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198968.2 | NP_001185897.1 | 6 | 695 | ||
| NM_001198969.2 | NP_001185898.1 | 6 | 695 | |||
| NM_011365.4 | NP_035495.2 | 6 | 695 | |||
| GenBank mRNA | AK029325.1 | - | 6 | 642 | ||
| AK034367.1 | - | 6 | 696 | |||
| AK042449.1 | - | 6 | 665 | |||
| AK122480.1 | - | 1 | 82 | |||
| AK163807.1 | - | 6 | 669 | |||
| BC138262.1 | - | 6 | 672 | 8861281 | ||
| BC171950.1 | - | 6 | 672 | 9054236 | ||

Itsn2
Genomic Map