siRNA Details
| Gene Symbol: Itsn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198968.2 | NP_001185897.1 | 3 | 329 | ||
| NM_001198969.2 | NP_001185898.1 | 3 | 329 | |||
| NM_011365.4 | NP_035495.2 | 3 | 329 | |||
| GenBank mRNA | AF132479.1 | - | 2 | 69 | ||
| AF132480.1 | - | 2 | 69 | |||
| AK029325.1 | - | 3 | 276 | |||
| AK034367.1 | - | 3 | 330 | |||
| AK042449.1 | - | 3 | 299 | |||
| AK163807.1 | - | 3 | 303 | |||
| BC138262.1 | - | 3 | 306 | 8861281 | ||
| BC171950.1 | - | 3 | 306 | 9054236 | ||

Itsn2
Genomic Map