siRNA Details
| Gene Symbol: Shcbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011369.2 | NP_035499.1 | 9, 10 | 1356 | ||
| GenBank mRNA | AF017152.1 | - | 9, 10 | 1356 | ||
| AK135226.1 | - | 1, 2 | 1978 | |||
| AK137496.1 | - | 8, 9 | 1838 | |||
| AK144966.1 | - | 9, 10 | 1379 | |||
| AK162943.1 | - | 9, 10 | 1370 | |||
| BC070455.1 | - | 9, 10 | 1376 | 30606785 | ||

Shcbp1
Genomic Map