siRNA Details
| Gene Symbol: Shfm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009169.2 | NP_033195.1 | 3 | 390 | ||
| GenBank mRNA | AK135462.1 | - | 343 | |||
| AK160432.1 | - | 3 | 383 | |||
| AK161690.1 | - | 3 | 363 | |||
| AK167914.1 | - | 3 | 340 | |||
| BC023490.1 | - | 3 | 347 | 5251089 | ||
| BC058117.1 | - | 3 | 356 | 6412144 | ||
| U41626.1 | - | 3 | 332 | |||
| X83589.1 | - | 3 | 365 | |||

Shfm1
Genomic Map