siRNA Details
| Gene Symbol: Shh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009170.3 | NP_033196.1 | 1 | 556 | ||
| GenBank mRNA | AK077688.1 | - | 1 | 557 | ||
| BC063087.1 | - | 1 | 543 | 6516263 | ||
| EU664592.1 | - | 1 | 102 | |||
| X76290.1 | - | 1 | 218 | |||

Shh
Genomic Map