siRNA Details
| Gene Symbol: Siah1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009172.2 | NP_033198.1 | 2 | 1165 | ||
| GenBank mRNA | BC046317.1 | - | 2 | 1165 | 6314641 | |
| Z19579.1 | - | 2 | 1093 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_080905, XM_006255211, XM_006255213, XM_008772345, AF389476, and others.

Siah1a
Genomic Map