siRNA Details
| Gene Symbol: IL13RA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001560.2 | NP_001551.1 | 3 | 291 | ||
| XM_011531336.1 | XP_011529638.1 | 3 | 569 | |||
| GenBank mRNA | AB209849.1 | - | 3 | 246 | ||
| AK313467.1 | - | 3 | 280 | |||
| BC009960.2 | - | 3 | 278 | 4300487 | ||
| BC015768.1 | - | 3 | 295 | 4868206 | ||
| U81379.3 | - | 3 | 288 | |||
| U81380.2 | - | 3 | 288 | |||
| Y09328.1 | - | 3 | 281 | |||
| Y10659.1 | - | 3 | 291 | |||

IL13RA1
Genomic Map