siRNA Details
| Gene Symbol: St6gal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252505.1 | NP_001239434.1 | 8 | 1649 | ||
| NM_001252506.1 | NP_001239435.1 | 7 | 1516 | |||
| NM_145933.4 | NP_666045.1 | 8 | 1980 | |||
| GenBank mRNA | AK034768.1 | - | 2 | 326 | ||
| AK084124.1 | - | 8 | 1815 | |||
| BC027833.1 | - | 5 | 1177 | 5321763 | ||
| BC092222.1 | - | 8 | 1958 | 6844069 | ||
| BC096026.1 | - | 8 | 1628 | 6833540 | ||
| D16106.1 | - | 7 | 1467 | |||

St6gal1
Genomic Map