siRNA Details
| Gene Symbol: St3gal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009179.3 | NP_033205.2 | 4 | 1787 | ||
| GenBank mRNA | AK029246.1 | - | 4 | 1702 | ||
| AK031052.1 | - | 4 | 1730 | |||
| AK034863.1 | - | 4 | 1728 | |||
| AK053827.1 | - | 4 | 1726 | |||
| AK131653.1 | - | 3 | 610 | |||
| BC015264.1 | - | 3 | 1029 | 4211307 | ||
| BC066064.1 | - | 4 | 1686 | 6850583 | ||
| X76989.1 | - | 3 | 693 | |||

St3gal2
Genomic Map