siRNA Details
| Gene Symbol: St6galnac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009180.3 | NP_033206.2 | 9 | 1100 | ||
| GenBank mRNA | AK004613.1 | - | 9 | 1101 | ||
| AK079216.1 | - | 1258 | ||||
| AK136381.1 | - | 1103 | ||||
| BC003276.1 | - | 9 | 2002 | |||
| BC010208.1 | - | 1052 | ||||
| BC023076.1 | - | 3 | 1548 | |||
| EU937514.1 | - | 3 | 1547 | |||
| X93999.1 | - | 9 | 1064 | |||

St6galnac2
Genomic Map