siRNA Details
| Gene Symbol: St6galnac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009180.3 | NP_033206.2 | 7 | 887 | ||
| GenBank mRNA | AK004613.1 | - | 7 | 888 | ||
| AK136381.1 | - | 890 | ||||
| BC003276.1 | - | 7 | 1789 | |||
| BC010208.1 | - | 839 | ||||
| BC023076.1 | - | 1 | 1335 | |||
| EU937514.1 | - | 1 | 1334 | |||
| X93999.1 | - | 7 | 851 | |||

St6galnac2
Genomic Map