siRNA Details
| Gene Symbol: Stil | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009185.3 | NP_033211.2 | 5 | 576 | ||
| GenBank mRNA | AK049223.1 | - | 5 | 513 | ||
| AK049550.1 | - | 5 | 513 | |||
| BC049865.1 | - | 5 | 530 | |||
| BC060706.1 | - | 5 | 491 | |||
| BC087545.1 | - | 5 | 515 | |||
| BC108373.1 | - | 5 | 492 | |||
| BC141175.1 | - | 5 | 449 | 9055787 | ||
| BC145493.1 | - | 5 | 449 | 9054056 | ||
| U36778.1 | - | 5 | 494 | |||

Stil
Genomic Map