siRNA Details
Gene Symbol: Six5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011383.1 | NP_035513.1 | 2 | 1033 | ||
GenBank mRNA | D83146.1 | - | 2 | 784 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Six5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011383.1 | NP_035513.1 | 2 | 1033 | ||
GenBank mRNA | D83146.1 | - | 2 | 784 |