siRNA Details
| Gene Symbol: Six5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011383.1 | NP_035513.1 | 2 | 1083 | ||
| GenBank mRNA | AK216841.1 | - | 1 | 22 | ||
| D83146.1 | - | 2 | 834 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Six5| Gene Symbol: Six5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011383.1 | NP_035513.1 | 2 | 1083 | ||
| GenBank mRNA | AK216841.1 | - | 1 | 22 | ||
| D83146.1 | - | 2 | 834 | |||